Description
- Acts as an intermediate piece between the plastic olive and the glass thread
- Enables a leak-proof, leakage-protected connection of components
- Prevents unwanted mass transfer in laboratory assemblies
- Compatible with DWK Life Sciences plastic olive and glass thread fittings
Specifications
Specifications
| For Use With (Equipment) | Plastic Hose Connections (Cat. No. 29 25506 03 and 29 247 05 04) |
| Quantity | 10/Pk. |
| Product Type | O-Ring |
| Material | Silicone (VMQ) |
| Diameter (Metric) Outside | 10mm |
| Autoclavable | Autoclavable |