Learn More
Grainger Replacement Sealing Ring (Gl32) PK10
Catalog No. 502749434
Encompass_Preferred
The Replacement Sealing Ring Gl32 is a replacement sealing ring for use with a variety of pumps and valves. It is made of Viton fluoroelastomer and has an inside diameter of 1.260 inches, an outside diameter of 1.575 inches, and a width of 0.172 inches. The maximum temperature rating is 400 degrees Fahrenheit. This Viton sealing ring can withstand a variety of chemicals and is designed to replace worn or damaged seals to help stop leaks. The 10-pack of replacement rings is manufactured in the USA by Replacement Parts Industries Inc.
Catalog No. 50-274-9434 Supplier Grainger Supplier No. 14N170
The Replacement Sealing Ring Gl32 is a replacement sealing ring for use with a variety of pumps and valves. It is made of Viton fluoroelastomer and has an inside diameter of 1.260 inches, an outside diameter of 1.575 inches, and a width of 0.172 inches. The maximum temperature rating is 400 degrees Fahrenheit. This Viton sealing ring can withstand a variety of chemicals and is designed to replace worn or damaged seals to help stop leaks. The 10-pack of replacement rings is manufactured in the USA by Replacement Parts Industries Inc.