Learn More
Grainger Replacement Sealing Ring (Gl45) PK10
Catalog No. 502749625
Encompass_Preferred
The Benchmark Scientific Sealing Ring 10-Pack is a set of 10 replacement sealing rings for Benchmark Scientific's line of glass media bottles. The rings are made from silicone and measure 38mm in diameter with a thickness of 3mm. They are designed to fit Benchmark Scientific's GL45 threaded media bottles in capacities ranging from 125mL to 2L. The silicone material provides a tight seal to prevent evaporation and contamination of media and solutions stored in the bottles. This 10-pack of sealing rings enables users to replace worn or damaged rings to maintain a secure seal on their Benchmark Scientific media bottles.
Catalog No. 50-274-9625 Supplier Grainger Supplier No. 14N168
The Benchmark Scientific Sealing Ring 10-Pack is a set of 10 replacement sealing rings for Benchmark Scientific's line of glass media bottles. The rings are made from silicone and measure 38mm in diameter with a thickness of 3mm. They are designed to fit Benchmark Scientific's GL45 threaded media bottles in capacities ranging from 125mL to 2L. The silicone material provides a tight seal to prevent evaporation and contamination of media and solutions stored in the bottles. This 10-pack of sealing rings enables users to replace worn or damaged rings to maintain a secure seal on their Benchmark Scientific media bottles.