Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Replacement Shelves

Catalog No. 11651152
Change view
Click to view available options
For Use With (Equipment):
3782 and 3782-1CE models
3790 and 3790-1CE
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
11-651-152 3790 and 3790-1CE
11-651-157 3782 and 3782-1CE models
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-651-152 Supplier Thermo Scientific™ Supplier No. 37908
Only null left
Add to Cart
Add to Cart

For Thermo Scientific™ Lab-Line™ Large-Capacity Freezers

TRUSTED_SUSTAINABILITY

Specifications

Type Shelf
Color White
For Use With (Equipment) 3790 and 3790-1CE
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.