Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Troemner™ Replacement Sleeves for Clamps

Catalog No. p-4677708 Shop All Troemner Products
Change view
Click to view available options
Size:
Small
Medium
Large
Material:
Fiberglass
Vinyl
Color:
Black
Red
For Use With (Equipment):
2-prong Clamps
3-prong Multipurpose Clamps
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Material Color For Use With (Equipment)
02-216-252 Medium Fiberglass Black 3-prong Multipurpose Clamps
02-216-245 Medium Vinyl Red 2-prong Clamps
02-216-253 Large Vinyl Red 3-prong Multipurpose Clamps
02-216-254 Large Fiberglass Black 3-prong Multipurpose Clamps
02-216-250 Small Fiberglass Black 3-prong Multipurpose Clamps
02-216-247 Large Vinyl Red 2-prong Clamps
02-216-251 Medium Vinyl Red 3-prong Multipurpose Clamps
02-216-246 Medium Fiberglass Black 2-prong Clamps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 02-216-252 Supplier Troemner™ Supplier No. 80862742
Only null left
Add to Cart
Add to Cart

Sleeves are easily removed for cleaning or replacement

  • Available in vinyl or fiberglass
  • Fiberglass sleeves are recommended for all applications above 100°C (212°F)
TRUSTED_SUSTAINABILITY

Specifications

Quantity 3/Pk.
Type Replacement Sleeve
Material Fiberglass
For Use With (Equipment) 3-prong Multipurpose Clamps
Length (Metric) 5 to 7 cm
Size Medium
Color Black
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.