Learn More
World Precision Instrument Replacement Sleeves for ISO-NOP
SDP
Catalog No. 502521253
Encompass
Keep your ISO-NOP sensor performing properly
The replacement membrane sleeves fit the 2mm (OD) ISO-NOP nitric oxide sensor. They are stainless steel tubes with an NO permeable membrane, and they come in a package of 4 sleeves.
Catalog No. 50-252-1253 Supplier World Precision Instrument Supplier No. 5436
Keep your ISO-NOP sensor performing properly
The replacement membrane sleeves fit the 2mm (OD) ISO-NOP nitric oxide sensor. They are stainless steel tubes with an NO permeable membrane, and they come in a package of 4 sleeves.