Learn More
Electron Microscopy Sciences Glass Sample Bottle Replacement Snap Cap, 22 ml
SDP
Catalog No. 5026141
Encompass_Preferred
For collecting and exhibiting a wide variety of specimens in the lab or field. Caps snap over a special sealing ring to form an effective barrier to moisture, making these bottles especially suitable for hydroscopic materials.
Capacity: 22 mm
Catalog No. 50-261-41 Supplier Electron Microscopy Sciences Supplier No. 6425122
For collecting and exhibiting a wide variety of specimens in the lab or field. Caps snap over a special sealing ring to form an effective barrier to moisture, making these bottles especially suitable for hydroscopic materials.
Capacity: 22 mm