Learn More
Analytical Sales & Services Replacement Socket Head Screws for 96 Position Radiochemistry Reactor
Catalog No. 5000804932
Encompass
Replacement Socket Head Screws for 96 Position Radiochemistry Reactor\n\n• 6-32 x 7/8” Long\n• Stainless Steel\n• Used to secure Top Cover to Transfer Plate and Base Vial Rack and provides compression sealing to Cap Mat/vials\n• Use with Radiochemistry Reactors (96950) or Conversion Kit (96940)\n\nQty/Pkg: 100
Catalog No. 50-008-04932 Supplier Analytical Sales & Services Supplier No. SHS152
Replacement Socket Head Screws for 96 Position Radiochemistry Reactor\n\n• 6-32 x 7/8” Long\n• Stainless Steel\n• Used to secure Top Cover to Transfer Plate and Base Vial Rack and provides compression sealing to Cap Mat/vials\n• Use with Radiochemistry Reactors (96950) or Conversion Kit (96940)\n\nQty/Pkg: 100