Learn More
MSC Buyers Products Replacement Spinner Motor, for SaltDogg SHPE Series Spreaders
Catalog No. 502835925
Encompass
This replacement spinner motor is designed for use with a variety of SaltDogg SHPE Series spreaders. It serves as the motor component for the spinner gearbox assembly, ensuring proper operation of the spreader's dispensing mechanism.
  • Designed for SHPE series spreaders
  • Compatible with electric auger models from 0.4 to 2.0 cubic yards
  • Fits models with extended chutes
  • Essential component for spinner gearbox operation
Catalog No. 50-283-5925 Supplier MSC Supplier No. 43235746
This replacement spinner motor is designed for use with a variety of SaltDogg SHPE Series spreaders. It serves as the motor component for the spinner gearbox assembly, ensuring proper operation of the spreader's dispensing mechanism.
  • Designed for SHPE series spreaders
  • Compatible with electric auger models from 0.4 to 2.0 cubic yards
  • Fits models with extended chutes
  • Essential component for spinner gearbox operation