Learn More
Analytical Sales & Services Replacement Stator for 2 Position, 10 Port High Pressure PEEK Switching Valve
Catalog No. 5000806395
Encompass
Stainless Steel Replacement Stator for 2 Position, 10 Port High Pressure PEEK Switching Valve (VXV7738-000)
Catalog No. 50-008-06395 Supplier Analytical Sales & Services Supplier No. 77738
Stainless Steel Replacement Stator for 2 Position, 10 Port High Pressure PEEK Switching Valve (VXV7738-000)