Learn More
Research Products International Corp Replacement Stainless Steel Chamber for Beadbeater
SDP
Catalog No. 501977578
Encompass
A BeadBeater large chamber made of stainless steel. The metal walls offer much better heat transfer than walls of the standard polycarbonate large chamber.
Net capacity is 350ml with a working volume of 150-200 ml of cell suspension. Strongly recommended for preparations requiring superior cooling during the homogenization process. This chamber is used with the standard rotor assembly and ice-water jacket included with a BeadBeater.
Catalog No. 50-197-7578 Supplier Research Products International Corp Supplier No. 141814
A BeadBeater large chamber made of stainless steel. The metal walls offer much better heat transfer than walls of the standard polycarbonate large chamber.
Net capacity is 350ml with a working volume of 150-200 ml of cell suspension. Strongly recommended for preparations requiring superior cooling during the homogenization process. This chamber is used with the standard rotor assembly and ice-water jacket included with a BeadBeater.