Learn More
Thermo Scientific™ Replacement Stopcock Assembly for Nalgene™ Separatory Funnels
GREENER_CHOICE
Catalog No. 14831126 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
Nalgene Separatory Funnels
Catalog No. For Use With (Equipment)
14-831-126 Nalgene Separatory Funnels
1 options
Catalog No. 14-831-126 Supplier Thermo Scientific™ Supplier No. 7143011050
  • $239.00
    $145.65 / Case of 1
    Save $93.35
    39% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Maintain separatory funnels with this Replacement Stopcock Assembly for Thermo Scientific™ Nalgene™ Separatory Funnels, which ensures a leakproof† fit without lubrication.
Maintain separatory funnels with this Replacement Stopcock Assembly for Thermo Scientific™ Nalgene™ Separatory Funnels, which ensures a leaktight fit without lubrication. This tetrafluoroethylene assembly has excellent chemical resistance and can be chemically disinfected.
  • For use with Nalgene™ Separatory Funnels made with Teflon™ fluoropolymer
  • Can be chemically sterilized
TRUSTED_SUSTAINABILITY
Specifications
Material Plastic
Description Replacement Stopcock Assembly; Stopcock Assy TFE 4301-2000 2 L
For Use With (Equipment) Nalgene Separatory Funnels
Quantity 1/Pk.; 1/Cs.
Teflon™ is a trademark of The Chemours Company FC, LLC used under license by Nalge Nunc International Corporation.