Learn More
Hach Company Replacement Stopper
Supplier:  Hach Company 2662300
Encompass
Replacement stopper for use with Gas washing bottle in the Hach Cold Vapor Mercury Apparatus Set (Prod No 2674400) to perform Cold Vapor Mercury Concentration Analysis Hach Method 10065
Catalog No. 50-002-57111