Learn More
Synthware™ Replacement for Straight Bore Glass Stopcock, Plug Only
GREENER_CHOICESDP
Catalog No. 31502029 Shop All Kemtech Products
Change view
Click to view available options
Plug Bore:
2 mm
4 mm
6 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Plug Bore
31-502-029 6 mm
31-502-027 2 mm
31-502-028 4 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 31-502-029 Supplier Synthware™ Supplier No. S462044
Add to Cart
Add to Cart
Glass plug ONLY.
Specifications
Type Replacement Stockcock
Plug Bore 6 mm
Material Borosilicate Glass
Autoclavable Autoclavable
For Use With (Application) Organic Synthesis
For Use With (Equipment) S69 Series
Plug Material Glass