Learn More
Synthware™ Replacement T-Bore Stopcock
GREENER_CHOICESDP
Catalog No. 31502014 Shop All Kemtech Products
Change view
Click to view available options
Plug Bore:
2 mm
4 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Plug Bore
31-502-014 4 mm
31-502-013 2 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 31-502-014 Supplier Synthware™ Supplier No. S201635
Add to Cart
Add to Cart
Plug has a T-shaped bore to connect any two or all three stems.
Specifications
Type T-Bore Stopcock
Plug Bore 4 mm
Material PTFE
For Use With (Application) Organic Synthesis
For Use With (Equipment) T-Bore Tap
Plug Material PTFE