Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Replacement Teflon O-rings

Catalog No. 116990100 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Packaging Quantity:
50 Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Quantity
11-699-0100 50 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-699-0100 Supplier MP Biomedicals™ Supplier No. 116990100
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

These Replacement Teflon O-rings are used in the cap of a SS Lysing Matrix tube to provide a leak-proof closure.

  • Replacement o-rings made of teflon designed to be inserted in the cap of the metal Lysing Matrix tubes to provide a leak-proof closure.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) 2 mL Metal SS Lysing Matrix Tube
Packaging Quantity 50 Pk.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.