Learn More
BRANDTECH™ BRAND™ Replacement Tip Cones and O-Rings for Transferpette™ S, Transferpette™ Electronic, and Transferpette™ Pro Multichannel Pipettes
Catalog No. 13689221 Shop All BRANDTECH Scientific Products
Change view
Click to view available options
For Use With (Equipment):
Electronic Pipette, 1 to 20 μL
Electronic Pipette, 20 μL
Multichannel Pipette, 0.5 to 10 μL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
13-689-221 Electronic Pipette, 1 to 20 μL
13-689-222 Electronic Pipette, 20 μL
13-688-677 Multichannel Pipette, 0.5 to 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 13-689-221 Supplier BRANDTECH™ BRAND™ Supplier No. 705678
Add to Cart
Add to Cart
Spare parts for Transferpette™ S, Transferpette™ electronic, and Transferpette™ pro, multichannel pipettes.
  • 705677: Includes two replacement tip cones for 0.5 to 10 μL nominal capacity Transferpette™ S, Transferpette™ electronic, and Transferpette™ pro pipettes with dual O-ring design, complete with mounted O-rings
  • 705678: Includes two replacement tip cones for 1 to 20 μL nominal capacity Transferpette™ electronic pipettes with dual O-ring design, complete with mounted O-rings
  • 705680: Includes two replacement tip cones compatible with older models of BRAND Transferpette™ electronic 1 to 20 μL multichannel pipettes with a single O-ring design
  • 703380: Includes 24 O-rings; designed for use with Transferpette™ S, Transferpette™ electronic, and Transferpette™ pro multichannel pipettes with nominal capacities of 0.5 to 10 μL and 1 to 20 μL
TRUSTED_SUSTAINABILITY
Specifications
Description Replacement tip cones for BRAND Transferpette S and electronic -8/12 multichannel pipettes, 20uL nominal capacity, 2-ring design
For Use With Transferpette Electronic
Material Polypropylene
Model Transferpette -8/12
Sterility Non-sterile
Type O-ring Kit
Volume (Metric) 20 μL
For Use With (Equipment) Electronic Pipette, 1 to 20 μL
Includes Nose cones, O-rings, seals, instructions
Quantity 2 Pack
WARNING: Reproductive Harm - www.P65Warnings.ca.gov