Learn More
Description
- formance
- Used for marking hard surfaces such as metal, glass, and plastic
- Construction
- Retractable tip
- Carbide or diamond tips
- Materials
- Carbide
- Diamond
Specifications
Specifications
| Tip Style | Retractable |
| Product Type | Replacement Tip |
| Color | Silver |
| Autoclavable | Autoclavable |
| Sterility | Non-sterile |
| Tip Material | Diamond |
| Latex Free | No |
| For Use With (Equipment) | Retractable Tip Evidence Scriber |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
