Learn More
Fine Science Tools REPLACEMENT TIP S & F 10/PK
SDP
Catalog No. NC9815046
Encompass
Product: Straight Fine Tips for Low Cost Cauterizer/ Tip Shape : Straight Fine/ Tip Diameter : 0.25mm/ Alloy : Stainless Steel/ Unit : 10
Catalog No. NC9815046 Supplier Fine Science Tools Supplier No. 1801002
Product: Straight Fine Tips for Low Cost Cauterizer/ Tip Shape : Straight Fine/ Tip Diameter : 0.25mm/ Alloy : Stainless Steel/ Unit : 10