Learn More
World Precision Instrument Replacement Tips for Reusable Rapid Biopsy Punch-504641
SDP
Catalog No. 502539321
Encompass
Stainless steel replacement tips for use with reusable rapid biopsy punch. Designed for heavy, numerous and precise sampling operations up to 2000 punches.
Catalog No. 50-253-9321 Supplier World Precision Instrument Supplier No. 504641
Stainless steel replacement tips for use with reusable rapid biopsy punch. Designed for heavy, numerous and precise sampling operations up to 2000 punches.