Learn More
Electron Microscopy Sciences Replacement Triangular Scraper
SDP
Catalog No. 50996717
Encompass
Hexagonal anodized aluminum handles with hardened tool steel tips that can be sharpend or pointed. OAL is 140 mm.
Catalog No. 50-996-717 Supplier Electron Microscopy Sciences Supplier No. 6209714
Hexagonal anodized aluminum handles with hardened tool steel tips that can be sharpend or pointed. OAL is 140 mm.