Learn More
Analytical Sales & Services Replacement UHMWPE Piston Seal
Catalog No. 5000805017
Encompass
Replacement UHMWPE Piston Seals for Shimadzu
Models LC-600 LC-9A LC-10AD LC-10ADvp LC-20AD LC-2010 & LC-20AB;
OEM Nos. 228-21975-91 & 228-32628-01
Qty/Pkg: 10PK
Catalog No. 50-008-05017 Supplier Analytical Sales & Services Supplier No. ADV99620
Replacement UHMWPE Piston Seals for Shimadzu
Models LC-600 LC-9A LC-10AD LC-10ADvp LC-20AD LC-2010 & LC-20AB;
OEM Nos. 228-21975-91 & 228-32628-01
Qty/Pkg: 10PK