Learn More
Perkin Elmer US LLC Replacement Upper Die for Sealer
Catalog No. 502111993
Encompass_Preferred
The Volatile Sample Pan Sealer is used to seal volatile, viscous, non-corrosive samples amd can be used in either Thermal Analysis or Elemental Analysis investigations.
Catalog No. 50-211-1993 Supplier Perkin Elmer US LLC Supplier No. 02191296
The Volatile Sample Pan Sealer is used to seal volatile, viscous, non-corrosive samples amd can be used in either Thermal Analysis or Elemental Analysis investigations.