Learn More
MSC Albion XP X-treme Plus Caster Wheel, Solid Polyurethane, 8 in., 1 ea.
Catalog No. 5000516005
Encompass
This solid polyurethane caster wheel is designed for demanding industrial applications requiring high load capacity and smooth mobility. The one-piece construction ensures durability, while the precision ball bearing system provides extremely low rolling resistance for easy transport. Its non-marking tread is engineered for excellent floor protection and reliable performance in a variety of material handling environments.
  • Wheel diameter of 8 in. with a 2 in. tread width
  • Hardness rating of 60 Shore D
  • Load capacity of 2,000 lb.
  • Precision ball bearing with 1/2 in. axle size
  • Temperature range of -40°F to +180°F
  • Non-marking crowned tread
  • Solid polyurethane material with a plastic core
  • Hub length of 2-7/16 in.
Catalog No. 50-005-16005 Supplier MSC Supplier No. 80126154
This solid polyurethane caster wheel is designed for demanding industrial applications requiring high load capacity and smooth mobility. The one-piece construction ensures durability, while the precision ball bearing system provides extremely low rolling resistance for easy transport. Its non-marking tread is engineered for excellent floor protection and reliable performance in a variety of material handling environments.
  • Wheel diameter of 8 in. with a 2 in. tread width
  • Hardness rating of 60 Shore D
  • Load capacity of 2,000 lb.
  • Precision ball bearing with 1/2 in. axle size
  • Temperature range of -40°F to +180°F
  • Non-marking crowned tread
  • Solid polyurethane material with a plastic core
  • Hub length of 2-7/16 in.