Learn More
Hach Company Replacement Silicone Wiper for Mechanical Cleaning Unit - For TU5300sc and TU5400sc Turbidimeters
Catalog No. NC1953260 Shop All Hach Company Products
Encompass
Replacement silicone wiper designed for the mechanical cleaning unit (automatic cleaning module) used on TU5-series turbidimeters. The wiper removes residue from vial surfaces to help maintain accurate turbidity measurements and should be replaced periodically based on sample conditions.
  • Silicone material for effective residue removal.
  • Compatible with TU5-series mechanical cleaning modules.
  • Recommended replacement every six months or as needed depending on sample type.
  • Helps maintain measurement accuracy by keeping vials clean.
  • Simple replacement procedure to minimize instrument downtime.
Catalog No. NC1953260 Supplier Hach Company Supplier No. LZY915
Replacement silicone wiper designed for the mechanical cleaning unit (automatic cleaning module) used on TU5-series turbidimeters. The wiper removes residue from vial surfaces to help maintain accurate turbidity measurements and should be replaced periodically based on sample conditions.
  • Silicone material for effective residue removal.
  • Compatible with TU5-series mechanical cleaning modules.
  • Recommended replacement every six months or as needed depending on sample type.
  • Helps maintain measurement accuracy by keeping vials clean.
  • Simple replacement procedure to minimize instrument downtime.