Learn More
Stoelting™ Replacement Wound Closure Clips
SDP
Catalog No. 10001025 Shop All Stoelting Co Products
Change view
Click to view available options
Model:
Replacement
Catalog No. Model
10-001-025 Replacement
1 options
Catalog No. 10-001-025 Supplier Stoelting™ Supplier No. 59035
Add to Cart
Add to Cart
Stoelting™ Replacement Wound Clips are individually packaged in heat sealed, peelaway envelopes for cleanliness and ease of handling and storage
  • Packaged 20 on a rack, with 10 racks per box (200 total), or 50 racks per case (1000 total)
  • Pyrogen-free
  • Non-sterile
  • Autoclavable plastic packaging allows sterilization of the clips without removing them from the packaging
Specifications
For Use With (Application) Neuroscience
Size 7 mm
Model Replacement
Includes Box of 100