Learn More
Analytical Sales & Services Replacements Thumb Screws for 96 Position Radiochemistry Reactor
Catalog No. 5000806057
Encompass
Replacements Thumb Screws for 96 Position Radiochemistry Reactor\n\n• M4-0.70mm x 5mm Long\n• Stainless Steel\n• For safe lifting/handling of Top Cover and Transfer Plate\n• Use with Radiochemistry Reactors (96950) or Conversion Kit (96940)\n\nQty/Pkg: Each
Catalog No. 50-008-06057 Supplier Analytical Sales & Services Supplier No. TS350
Add to Cart
Add to Cart
Replacements Thumb Screws for 96 Position Radiochemistry Reactor\n\n• M4-0.70mm x 5mm Long\n• Stainless Steel\n• For safe lifting/handling of Top Cover and Transfer Plate\n• Use with Radiochemistry Reactors (96950) or Conversion Kit (96940)\n\nQty/Pkg: Each