Learn More
Mitegen Replacement Polycarbonate Washers, For Securing Ring Magnets in Universal V1-Pucks & SSRL Cassettes, 100/Pack
SDP
Catalog No. NC3659625
Encompass
These high-quality Replacement Polycarbonate Washers are essential components for maintaining the secure functionality of your Universal V1-Pucks and SSRL Sample Storage Cassettes. Designed specifically to hold ring magnets firmly in place within the sample ports, these washers play a critical role in preventing sample loss and ensuring sample integrity during cryogenic storage and handling. Crafted from durable polycarbonate, they offer excellent resistance to extreme temperatures and provide reliable performance in demanding laboratory environments. As removable components, they facilitate easy replacement as needed, supporting efficient routine maintenance. This 100-pack provides an ample supply of washers, ensuring continuous and secure sample management for various research applications. Consistent replacement of these washers is key to safeguarding valuable samples and optimizing the longevity of your cryogenic storage system.
Catalog No. NC3659625 Supplier Mitegen Supplier No. MCP111007100
These high-quality Replacement Polycarbonate Washers are essential components for maintaining the secure functionality of your Universal V1-Pucks and SSRL Sample Storage Cassettes. Designed specifically to hold ring magnets firmly in place within the sample ports, these washers play a critical role in preventing sample loss and ensuring sample integrity during cryogenic storage and handling. Crafted from durable polycarbonate, they offer excellent resistance to extreme temperatures and provide reliable performance in demanding laboratory environments. As removable components, they facilitate easy replacement as needed, supporting efficient routine maintenance. This 100-pack provides an ample supply of washers, ensuring continuous and secure sample management for various research applications. Consistent replacement of these washers is key to safeguarding valuable samples and optimizing the longevity of your cryogenic storage system.