Learn More
Mitegen Replacement Ring Magnets, Nickel Coated NdFeB, 10.4mm OD x 7.6mm ID x 2.5mm, For Universal V1-Pucks & SSRL Cassettes, 100/Pack
SDP
Catalog No. NC3659627
Encompass
These high-quality Replacement Ring Magnets are essential for maintaining the integrity and functionality of your Universal V1-Pucks and SSRL Sample Storage Cassettes. Each magnet is crafted from durable nickel-coated NdFeB (Neodymium-Iron-Boron), ensuring strong and reliable magnetic force for securely holding samples in their designated ports. Measuring precisely 10.4 mm outer diameter, 7.6 mm inner diameter, and 2.5 mm thickness, these ring magnets provide an exact fit for compatible equipment. They are designed to be held firmly in place by removable polycarbonate washers, enabling easy replacement as needed. Ensuring proper magnetic strength is crucial for preventing sample loss and maintaining sample integrity within cryogenic environments. This 100-pack provides ample supply for routine maintenance, supporting continuous and secure sample management in various laboratory settings.
Catalog No. NC3659627 Supplier Mitegen Supplier No. MCP111006100
These high-quality Replacement Ring Magnets are essential for maintaining the integrity and functionality of your Universal V1-Pucks and SSRL Sample Storage Cassettes. Each magnet is crafted from durable nickel-coated NdFeB (Neodymium-Iron-Boron), ensuring strong and reliable magnetic force for securely holding samples in their designated ports. Measuring precisely 10.4 mm outer diameter, 7.6 mm inner diameter, and 2.5 mm thickness, these ring magnets provide an exact fit for compatible equipment. They are designed to be held firmly in place by removable polycarbonate washers, enabling easy replacement as needed. Ensuring proper magnetic strength is crucial for preventing sample loss and maintaining sample integrity within cryogenic environments. This 100-pack provides ample supply for routine maintenance, supporting continuous and secure sample management in various laboratory settings.