Learn More
Microsolv Technology Corporation Performance Qualification HPLC Column. Replacement, for use with the HSQ Kit. Includes a Certificate of Performance. 1 EA
Supplier:  Microsolv Technology Corporation 4910175C
Encompass_Preferred
This specially selected HPLC Column is tested and certified by an independent testing laboratory for specific performance in Performance Qualification.Replacement for the original column supplied with the MicroSolv HSQ (High Speed Qualification) Kit, it is 75mm long and 4.6mm ID. The particle size of the Column phase is 5um.
Catalog No. 50-226-9765