Learn More
Microsolv Technology Corporation Performance Qualification HPLC Column. Replacement, for use with the PQ Kit. Includes a Certificate of Performance. 1 EA
Catalog No. 502269764
Encompass_Preferred
This specially selected HPLC Column is tested and certified by an independent testing laboratory for specific performance in Performance Qualification.Replacement for the original column supplied with the MicroSolv PQ (Performance Qualification) Kit, it is 150mm long and 4.6mm ID. The particle size of the Column phase is 5um.
This item is not returnable. View return policy
Catalog No. 50-226-9764 Supplier Microsolv Technology Corporation Supplier No. 4910150C
This specially selected HPLC Column is tested and certified by an independent testing laboratory for specific performance in Performance Qualification.Replacement for the original column supplied with the MicroSolv PQ (Performance Qualification) Kit, it is 150mm long and 4.6mm ID. The particle size of the Column phase is 5um.