Learn More
Microsolv Technology Corporation Ferrules, Replacement, for Di-Ad, Direct Adaptive HPLC Column Connectors. PEEK. 10/PK.
Catalog No. 502269783
Encompass_Preferred
After several hundred connections to HPLC columns or to the instruments, it is advisable to replace the "all PEEK Ferrule" of a Direct Adaptive (Di-Ad™) HPLC column connector, to insure good connections and prevent leaking during use. It is easy and simple to remove and insert this replacement.
This item is not returnable. View return policy
Catalog No. 50-226-9783 Supplier Microsolv Technology Corporation Supplier No. 4991101
After several hundred connections to HPLC columns or to the instruments, it is advisable to replace the "all PEEK Ferrule" of a Direct Adaptive (Di-Ad™) HPLC column connector, to insure good connections and prevent leaking during use. It is easy and simple to remove and insert this replacement.