Learn More
Vici Precision Sampling, Inc. Replacement Cutting Wheel
Catalog No. NC0418533
Encompass_Preferred
This is the ideal tool for cutting 1/16" and 1/8" stainless steel tubing, leaving smooth, uniform cuts requiring a minimum of deburring and/or reaming. The easily replaced cutting wheel scores tubing, leaving cuts which allow tube-to-tube and tube-to-frit connections.
Catalog No. NC0418533 Supplier Vici Precision Sampling, Inc. Supplier No. JR793
This is the ideal tool for cutting 1/16" and 1/8" stainless steel tubing, leaving smooth, uniform cuts requiring a minimum of deburring and/or reaming. The easily replaced cutting wheel scores tubing, leaving cuts which allow tube-to-tube and tube-to-frit connections.