Learn More
Sigma Aldrich Fine Chemicals Biosciences Replica plater for 96 well1EA
Catalog No. 502904721
Encompass
The prongs are 3/4 inch long and 0.125 inches in diameter. Each device has a semi-circular prong in the upper left corner for unambiguous orientation of the replica plate. All other prongs are circular.
Catalog No. 50-290-4721 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. R23831EA
The prongs are 3/4 inch long and 0.125 inches in diameter. Each device has a semi-circular prong in the upper left corner for unambiguous orientation of the replica plate. All other prongs are circular.