Learn More
Sunrise Science Products Velvet Replica Plating Pads, 9×9 (10 pack)
Catalog No. NC2885603
Encompass
Use with 150 mm Replica Plating Apparatus (#3005-002) to transfer colonies while retaining colony orientation.
Catalog No. NC2885603 Supplier Sunrise Science Products Supplier No. 3005012
Use with 150 mm Replica Plating Apparatus (#3005-002) to transfer colonies while retaining colony orientation.