Learn More
VWR International repliQa HiFi ToughMix, 100 reactions
Supplier:  VWR International 76382788
Encompass_Preferred
Superior speed and inhibitor tolerance.High fidelity with >90× wild type TaqUp to 3× faster PCR results with extension rates as fast as 1 kb/secTough tested and tolerant to a wide range of PCR inhibitorsSuperior sensitivity means higher yields from lower inputs as little as 2 pgHas the ability to amplify long fragments of genomic DNA (+24 kb) and lambda DNA (+40 kb)The repliQa HiFi ToughMix is a 2×, ready-to-use master mix that contains all the components for high fidelity PCR amplification, including a genetically modified DNA polymerase coupled with hot start antibodies. This unique, next generation master mix provides 90× higher fidelity compared to Taq, while reducing time to PCR results by 2 to 3×. The extreme speed is enabled by extension times as fast as 1 to 10 kb/sec depending on target length. The enzyme is coupled with the industry leading ToughMix which is tolerant to a wide variety of inhibitors.
Catalog No. NC3591680