Learn More
Molecular Devices Molecular Devices dsDNA Assay Kit, Explorer, 5 pg-3 ng
Catalog No. 5000014465
Encompass_Preferred
The SpectraMax Quant AccuBlue Pico dsDNA Assay Kit is a fluorescence-based assay for sensitive quantitation of double-stranded DNA in low-concentration samples. The Explorer kit provides reagents sufficient for two 96-well microplates and is optimized for use with microplate readers.
  • Sensitive fluorescence-based dsDNA quantitation from 5 pg to 3 ng.
  • Explorer kit format sufficient for two 96-well microplates.
  • Includes dsDNA-selective fluorescent dye and calibration standards.
  • Optimized for low-volume microplate assays and microplate readers.
Catalog No. 50-000-14465 Supplier Molecular Devices Supplier No. R8354
The SpectraMax Quant AccuBlue Pico dsDNA Assay Kit is a fluorescence-based assay for sensitive quantitation of double-stranded DNA in low-concentration samples. The Explorer kit provides reagents sufficient for two 96-well microplates and is optimized for use with microplate readers.
  • Sensitive fluorescence-based dsDNA quantitation from 5 pg to 3 ng.
  • Explorer kit format sufficient for two 96-well microplates.
  • Includes dsDNA-selective fluorescent dye and calibration standards.
  • Optimized for low-volume microplate assays and microplate readers.