Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Repositionable Vinyl Cloth Wire and Cable Labels

Catalog No. 19129815 Shop All Brady Corporation Products
Change view
Click to view available options
Dimensions (L x W):
1 x 1 in.
Quantity:
160 Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W) Quantity
19-129-815 1 x 1 in. 160 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-129-815 Supplier Brady™ Supplier No. M49498

Brady™ Repositionable Vinyl Cloth Wire and Cable Labels features CleanLift™ B-498 Series Vinyl Cloth with strong permanent adhesive that is also repositionable if needed.

  • The adhesive and cloth backing offers excellent holding power, yet allows for clean removal and repositioning
  • Sticks to any clean, dry surface
  • Specially formulated topcoat for excellent print quality
  • All-in-one cartridge includes label supply and R6200 Series ribbon

Specifications

Product Type Label
Color Black on White
For Use With (Equipment) BMP41 Label Printer, BMP51 Label Printer, BMP53 Label Printer
Material Vinyl Cloth (B-498)
Print Technology Thermal Transfer
Dimensions (L x W) 1 x 1 in.
Adhesive Type Permanent Acrylic
Sterility Non-sterile
Finish Semi-gloss
Format Cartridge, Ribbon
Certifications/Compliance UL Recognized to UL969, RoHS Compliant to 2005/618/EC MCV
Quantity 160 Pk.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.