Learn More
Electron Microscopy Sciences Repositionable Dry Mounts, Size 8x10"
SDP
Catalog No. 5028336
Encompass_Preferred
RDM is a pressure-sensitive mounting material that consist of a microthin, clear Mylar film coated on both sides with an adhesive. It is specially formulated for photographic purposes. This "delayed-set" adhesive allows for repositioning of prints before providing a permanent bond. No heat is needed. The Mylar base will not absorb moisture or change its characteristics through time; thereby it eliminates the common mounting problems that are experienced with other mounting materials. RDM sheets are made 1/4" oversize which eliminates alignment problems and makes it easier to use
Catalog No. 50-283-36 Supplier Electron Microscopy Sciences Supplier No. 77604
RDM is a pressure-sensitive mounting material that consist of a microthin, clear Mylar film coated on both sides with an adhesive. It is specially formulated for photographic purposes. This "delayed-set" adhesive allows for repositioning of prints before providing a permanent bond. No heat is needed. The Mylar base will not absorb moisture or change its characteristics through time; thereby it eliminates the common mounting problems that are experienced with other mounting materials. RDM sheets are made 1/4" oversize which eliminates alignment problems and makes it easier to use