Learn More
TARGETMOL CHEMICALS INC REPOTRECTINIB 10MG
SDP
Catalog No. 502579974
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Repotrectinib (TPX-0005) is a potent ALK/ROS1/TRK inhibitor with IC50 of 1.01 nM 5.3 nM 1.08 nM and 1.26 nM for WT ALK SRC ALK L1196M and ALK G1202R respectively. purity: 99%
Catalog No. 50-257-9974 Supplier TARGETMOL CHEMICALS INC Supplier No. T407110MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Repotrectinib (TPX-0005) is a potent ALK/ROS1/TRK inhibitor with IC50 of 1.01 nM 5.3 nM 1.08 nM and 1.26 nM for WT ALK SRC ALK L1196M and ALK G1202R respectively. purity: 99%