Learn More
Lifeline Cell Technology ReproLife Media Kit
SDP
Catalog No. NC1944651
Encompass_Preferred
ReproLife Complete Kit. Culture medium for growth of female reproductive epithelial cells (vaginal, uterine, and fallopian tube epithelial cells). LM-0044 ReproLife Basal Medium and LS-1097 ReproLife LifeFactors Kit
Catalog No. NC1944651 Supplier Lifeline Cell Technology Supplier No. LL0068
ReproLife Complete Kit. Culture medium for growth of female reproductive epithelial cells (vaginal, uterine, and fallopian tube epithelial cells). LM-0044 ReproLife Basal Medium and LS-1097 ReproLife LifeFactors Kit