Learn More
STEMCELL Technologies Resazurin (Sodium Salt), 5 g
SDP
Catalog No. NC1451439
Encompass
ReproRNA™-OKSGM is a single-stranded RNA replicon vector that contains five reprogramming factors: OCT4, KLF-4, SOX2, GLIS1, and c-MYC, as well as a puromycin-resistance gene (Figure 1). This RNA vector reprograms somatic cells, such as fibroblasts, into induced pluripotent stem (iPS) cells with high efficiency and only requires a single transfection step (Figure 2). When used together with ReproTeSR™ reprogramming medium, the generation of iPS cell colonies can be achieved under feeder-free conditions with superior colony morphology and similar reprogramming efficiency to feeder-based systems (Figures 3 - 4). ReproRNA™-derived iPS cell colonies also express markers of undifferentiated cells and retain a normal karyotype (Figures 5-6). Subsequently, iPS cells generated with ReproRNA™-OKSGM can be maintained in TeSR™ maintenance media and further differentiated into cells of all three germ layers (Figure 7).
Catalog No. NC1451439 Supplier STEMCELL Technologies Supplier No. 05930
ReproRNA™-OKSGM is a single-stranded RNA replicon vector that contains five reprogramming factors: OCT4, KLF-4, SOX2, GLIS1, and c-MYC, as well as a puromycin-resistance gene (Figure 1). This RNA vector reprograms somatic cells, such as fibroblasts, into induced pluripotent stem (iPS) cells with high efficiency and only requires a single transfection step (Figure 2). When used together with ReproTeSR™ reprogramming medium, the generation of iPS cell colonies can be achieved under feeder-free conditions with superior colony morphology and similar reprogramming efficiency to feeder-based systems (Figures 3 - 4). ReproRNA™-derived iPS cell colonies also express markers of undifferentiated cells and retain a normal karyotype (Figures 5-6). Subsequently, iPS cells generated with ReproRNA™-OKSGM can be maintained in TeSR™ maintenance media and further differentiated into cells of all three germ layers (Figure 7).