Learn More
STEMCELL Technologies ReproTeSR™ Medium for Reprogramming (2-Component), Size: 500 mL
SDP
Catalog No. 501974366
Encompass
ReproTeSR™ (2-Component) is a complete and xeno-free reprogramming medium developed for generating human induced pluripotent stem (iPS) cells from fibroblasts, urine-derived cells, and blood-derived cells such as CD34+ or erythroid precursor cells under feeder-free conditions.ReproTeSR™ is also available as part of an integrated set of tools that are optimized for reprogramming blood-derived cell types expanded in vitro from peripheral blood, such as CD34+ cells (CD34+ Progenitor Reprogramming Kit, Catalog #05925) or erythroid cells (Erythroid Progenitor Reprogramming Kit, Catalog #05924).
Catalog No. 50-197-4366 Supplier STEMCELL Technologies Supplier No. 05926
ReproTeSR™ (2-Component) is a complete and xeno-free reprogramming medium developed for generating human induced pluripotent stem (iPS) cells from fibroblasts, urine-derived cells, and blood-derived cells such as CD34+ or erythroid precursor cells under feeder-free conditions.ReproTeSR™ is also available as part of an integrated set of tools that are optimized for reprogramming blood-derived cell types expanded in vitro from peripheral blood, such as CD34+ cells (CD34+ Progenitor Reprogramming Kit, Catalog #05925) or erythroid cells (Erythroid Progenitor Reprogramming Kit, Catalog #05924).