Learn More
Medchemexpress LLC Reproxalap 50mg | 916056-79-6 | 50MG
SDP
Catalog No. 5000239929
Encompass_Preferred
Reproxalap (ADX-102) is an active aldehyde sequestering agent Reproxalap reduces the PKC activity Reproxalap blocks caspase 3/7 activation Reproxalap protects cells from the cytotoxicity of C18 0-al Reproxalap has anti-inflammatory and pain-relieving effects Reproxalap is used in studies of dry eye allergic conjunctivitis and non-infectious anterior uveitis[1 [2 [3 [4 [5
Catalog No. 50-002-39929 Supplier Medchemexpress LLC Supplier No. HY10715050MG
Reproxalap (ADX-102) is an active aldehyde sequestering agent Reproxalap reduces the PKC activity Reproxalap blocks caspase 3/7 activation Reproxalap protects cells from the cytotoxicity of C18 0-al Reproxalap has anti-inflammatory and pain-relieving effects Reproxalap is used in studies of dry eye allergic conjunctivitis and non-infectious anterior uveitis[1 [2 [3 [4 [5