Learn More
Medchemexpress LLC Reproxalap | 916056-79-6 | MFCD28502442 | 98.5% | 236.70 g/mol | C12H13ClN2O | 10 MG
SDP
Catalog No. 5000239831
Encompass_Preferred
Reproxalap (ADX-102) is a small-molecule aldehyde-sequestering agent supplied as a 10 mg research reagent. It is used in preclinical studies targeting reactive aldehyde species and shows activity in assays related to PKCα inhibition and caspase 3/7 blockade; it has been investigated for ocular inflammatory conditions such as dry eye disease. For research use only.
Catalog No. 50-002-39831 Supplier Medchemexpress LLC Supplier No. HY10715010MG
Reproxalap (ADX-102) is a small-molecule aldehyde-sequestering agent supplied as a 10 mg research reagent. It is used in preclinical studies targeting reactive aldehyde species and shows activity in assays related to PKCα inhibition and caspase 3/7 blockade; it has been investigated for ocular inflammatory conditions such as dry eye disease. For research use only.