Learn More
Medchemexpress LLC HY-13012 10mg Medchemexpress, RepSox CAS:446859-33-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1301210MG
Encompass_Preferred
Medchemexpress, HY-13012 10mg RepSox CAS:446859-33-2 RepSox is a potent and selective of the TGFβR-1/ALK5 inhibitor which inhibits ALK5 autophosphorylation with IC50 of 4 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8940