Learn More
Selleck Chemical LLC RepSox 10mg 446859-33-2 E-616452, SJN 2511,ALK5 Inhibitor II
SDP
Supplier:  Selleck Chemical LLC S722310MG
Encompass_Preferred
RepSox (E-616452, SJN 2511, ALK5 Inhibitor II) is a potent and selective inhibitor of the TGF?R-1/ALK5 with IC50 of 23 nM and 4 nM for ATP binding to ALK5 and ALK5 autophosphorylation in cell-free assays, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0951658