Learn More
STEMCELL Technologies RepSox (Hydrochloride), Size: 25 mg
SDP
Supplier:  STEMCELL Technologies 72394
Encompass
RepSox is a cell permeable, selective inhibitor of the TGF-β type 1 receptor (TGFβRI) ALK5 (IC₅₀ = 4, 18, and 23 nM for ALK5 autophosphorylation, TGF-β cellular assay, and ALK5 binding in HepG2 cells, respectively; Gellibert et al.). This inhibitor demonstrated less potent activity (IC₅₀ > 16 μM) against 9 related kinases, including p38 MAPK and GSK3 (Gellibert et al.). This product is supplied as the hydrochloride salt of the molecule.REPROGRAMMING· Enhances reprogramming of mouse embryonic fibroblasts (MEFs) that have been transduced with OCT4, KLF4, and c-MYC (Ichida et al.; Subramanyam et al.)· Direct lineage reprogramming of fibroblasts to mature neurons, in combination with CHIR99021, Valproic Acid, Forskolin, SP600125, Gö6983 and Y-27632 (Hu et al.).DIFFERENTIATION· Alone or in combination with forskolin, dexamethasone, and nicotinamide, induces differentiation of human pancreatic progenitor cells into insulin-producing cells (Kunisada et al.; Rezania et al.).
Catalog No. 50-197-5916