Description
Grease-free Solvent Still Head. Access port has threaded 14/10 joint.
Joint accepts:
- PTFE lined silicone septum/compression cap
- Standard 14/20 glass or PTFE stopper
Specifications
Specifications
| Includes | PTFE Stopper and Nylon Compression Cap |
| For Use With (Application) | Organic Synthesis |
| Type | Grease-free Repurification |
| Material | Borosilicate Glass |
| Capacity (Metric) | 250 mL |
| Quantity | 1 Ea. |
| Size | 24/40 |