Learn More
Synthware™ Repurification Solvent Still Head, Grease-free
GREENER_CHOICESDP
Catalog No. 31501849 Shop All Kemtech Products
Change view
Click to view available options
Capacity (Metric):
250 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
31-501-849 500 mL
31-501-848 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 31-501-849 Supplier Synthware™ Supplier No. S134500
Add to Cart
Add to Cart
Head only. Condenser and flasks not supplied.
Grease-free Solvent Still Head. Access port has threaded 14/10 joint.
Joint accepts:
  • PTFE lined silicone septum/compression cap
  • Standard 14/20 glass or PTFE stopper
Specifications
Includes PTFE Stopper and Nylon Compression Cap
For Use With (Application) Organic Synthesis
Type Grease-free Repurification
Material Borosilicate Glass
Capacity (Metric) 500 mL
Quantity 1 Ea.
Size 24/40